Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystathionase Antibody (OTI1E12), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270488MFV610
Description
Cystathionase Monoclonal antibody specifically detects Cystathionase in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Cystathionase | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Canine, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI1E12 | |
Flow Cytometry | |
cystathionase (cystathionine gamma-lyase), cystathionine gamma-lyase, cysteine desulfhydrase, EC 4.4.1, EC 4.4.1.1, gamma-cystathionase, homoserine deaminase, homoserine dehydratase, MGC9471 | |
Full length human recombinant protein of human CTH (NP_001893) produced in HEK293T cell. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
1491 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction