Learn More
Invitrogen™ Cystatin 2 Recombinant Rabbit Monoclonal Antibody (005)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530409
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Cystatin SA/CST2.
The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
Specifications
Cystatin 2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P09228 | |
CST2 | |
Recombinant Human Cystatin SA/CST2 Protein (Met1-Ala141). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
005 | |
Unconjugated | |
CST2 | |
CST2; cystatin 2; cystatin S5; cystatin SA; cystatin-2; Cystatin-S5; cystatin-SA; cysteine-proteinase inhibitor; salivary cysteine (thiol) protease inhibitor | |
Rabbit | |
Protein A | |
RUO | |
1470 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.