Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin-8 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP147219
Description
Cystatin-8 Polyclonal specifically detects Cystatin-8 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Cystatin-8 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CRESCystatin-related epididymal spermatogenic protein, cystatin 8 (cystatin-related epididymal specific), cystatin-8, cystatin-related epididymal-specific | |
Goat | |
Affinity Purified | |
RUO | |
10047 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_005483.1 | |
CST8 | |
Peptide with sequence C-DPKKNETGVLRK corresponding to internal region according to NP_005483.1. | |
0.1 mg | |
Primary | |
This antibody recognizes and epitope within the Internal region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction