Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin 9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556080 Supplier Invitrogen™ Supplier No. PA556080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGNNKIVQDP MFFATVEFAL NTFNVQSKEE HAYRLLRVLS SWREDSMDRK WRGKMVFSMN LQLRQTVCRK FEDDIDNCPF QESLELNNVR QGISFPQVHS CGCCMGCGV Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 39%.

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a secreted protein that may play a role in hematopoietic differentiation or inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin 9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CST9
Gene Accession No. Q5W186
Gene Alias CLM; CST9; CTES7A; cystatin 9; cystatin 9 (testatin); cystatin-9; Cystatin-like molecule; testatin
Gene Symbols CST9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cystatin 9. Recombinant protein control fragment (Product #RP-88587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.