Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody (1B12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259470F
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Cystatin A | |
Monoclonal | |
FITC | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
Recombinant human Cystatin A (1-98aa) purified from E. coli | |
0.1 mL | |
Cancer, Tumor Suppressors | |
1475 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence | |
1B12 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction