Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody (236), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289524PCP
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Western BlotSpecifications
Cystatin A | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
236 | |
Western Blot | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin A (Uniprot#: P01040; Ile2-Phe98). | |
0.1 mL | |
Cancer, Tumor Suppressors | |
1475 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction