Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody (CPTC-CSTA-1), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308498JF549
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Cystatin A | |
Monoclonal | |
Janelia Fluor 549 | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
Recombinant full-length human Cystatin A protein (Uniprot: P01040) | |
100 μg | |
Cancer, Tumor Suppressors | |
1475 | |
Store at 4°C in the dark. | |
IgG2c κ |
Western Blot, Immunohistochemistry (Paraffin) | |
CPTC-CSTA-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction