Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299327V
Description
Cystatin A Polyclonal specifically detects Cystatin A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cystatin A | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
Produced in rabbits immunized with purified, recombinant Cystatin A (P01040; Ile 2-Phe 98). | |
0.1 mL | |
Cancer, Tumor Suppressors | |
1475 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction