Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Cystatin A |
---|---|
Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cystatin A Polyclonal antibody specifically detects Cystatin A in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Cystatin A | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
0.2 um filtered solution in PBS | |
1475 | |
IgG | |
Antigen and protein A Affinity-purified |
ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
Produced in rabbits immunized with purified, recombinant Human Cystatin A (Uniprot#: P01040; Ile 2-Phe 98) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title