Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A Antibody (SR1147), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32227550UL
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Cystatin A | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1147 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
A synthesized peptide derived from human Cystatin A (Uniprot #: P01040) | |
50 μg | |
Cancer, Tumor Suppressors | |
1475 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction