Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cystatin A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551502
Description
Immunogen sequence: IPGGLSEAKP ATPEIQEIVD KVKPQLEEKT NETYGKLEAV QYKTQVVAGT NYYIKVRAGD NKYMHLKVFK SLPGQNEDLV Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 65%.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.
Specifications
Cystatin A | |
Polyclonal | |
Unconjugated | |
CSTA | |
AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P01040 | |
CSTA | |
Recombinant protein corresponding to Human Cystatin A. Recombinant protein control fragment (Product #RP-93681). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction