Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577929 Supplier Invitrogen™ Supplier No. PA577929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: THP-1, HL-60 Predicted Reactivity: Rhesus Monkey (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CSTA
Gene Accession No. P01040
Gene Alias AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFA
Gene Symbols CSTA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cystatin A. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.