Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596258 Supplier Invitrogen™ Supplier No. PA596258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMCGAPSATQ PATAETQHIA DQVRSQLEEK ENKKFPVFKA VSFKSQVVAG TNYFIKVHVG DEDFVHLRVF QSLPHENKPL TLSNYQTNKA KHDELTYF; Positive Samples: HepG2, SW480, DU145, A-549, A-431, MCF7; Cellular Location: Cytoplasm, Nucleus.

Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSTB
Gene Accession No. P01041, P04080
Gene Alias AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609)
Gene Symbols CSTB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human CSTB (NP_0000911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1476, 25308
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.