Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin B/Stefin B Mouse anti-Human, Clone: CPTC-CSTB-3, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Cystatin B/Stefin B
Clone CPTC-CSTB-3
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123050
SDP
View Documents
Novus Biologicals
NBP30778820UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cystatin B/Stefin B
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
1476
IgG1 κ
Protein A or G purified
CPTC-CSTB-3
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B
Recombinant full-length human Cystatin B/Stefin B protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.