Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin C Antibody (002), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290859V
Description
Cystatin C Monoclonal specifically detects Cystatin C in Rat samples. It is validated for ELISA.Specifications
Cystatin C | |
Monoclonal | |
DyLight 405 | |
ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Cystatin C (P14841-1; Met1-Ala140). | |
0.1 mL | |
Cancer, Epitope Tags, Signal Transduction | |
1471 | |
Rat | |
Purified |
ELISA | |
2 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse CST3 Human CST3 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction