Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin C Antibody (Cyst13), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105763AF532
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for ELISA, Immunoassay, ELISASpecifications
Cystatin C | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunoassay, ELISA | |
Cyst13 | |
ELISA, Immunoassay, Sandwich ELISA | |
ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine. | |
0.2 mL | |
Cancer, Epitope Tags, Signal Transduction | |
1471 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction