Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin C Antibody (Cyst19cc), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105766B
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for ELISA, Immunoassay, Sandwich ELISA.Specifications
Cystatin C | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CST3 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine. | |
0.2 mL | |
Cancer, Epitope Tags, Signal Transduction | |
Detects human Cystatin C, a member of the type 2 cystatin superfamily. Cystatin C is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsines), and pathogenic. | |
Store at 4°C in the dark. |
ELISA, Immunoassay | |
Cyst19cc | |
ELISA, Immunoassay, Sandwich ELISA | |
ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction