Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin C Antibody (Cyst23), Janelia Fluor™ 549

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062126JF549
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for Sandwich ELISA.Specifications
| Cystatin C | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CST3 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine. | |
| 0.2 mL | |
| Cancer, Epitope Tags, Signal Transduction | |
| NB100-62126 detects human Cystatin C, a member of the type 2 cystatin superfamily. Cystatin C is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsines), and pathogenic. | |
| Store at 4°C in the dark. |
| ELISA | |
| Cyst23 | |
| Sandwich ELISA | |
| ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction