Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin C Monoclonal Antibody (Cyst10)
GREENER_CHOICE

Catalog No. PIMA181926
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA181926 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA181926 Supplier Invitrogen™ Supplier No. MA181926
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Human Cystatin C antibody, clone Cyst10 detects human Cystatin C, a member of the type 2 cystatin superfamily.

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin C
Applications ELISA
Classification Monoclonal
Clone Cyst10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene Cst3
Gene Accession No. P01034
Gene Alias AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227
Gene Symbols Cst3
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1471
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.