Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589137 Supplier Invitrogen™ Supplier No. PA589137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPGKPPRLV GGPMDASVEE EGVRRALDFA VGEYNKASND MYHSRALQVV RARKQIVAGV NYFLDVELGR TTCTKTQPNL DNCPFHDQPH LKRKAFCSFQ IYAVPWQGTM TLSKSTCQDA; Positive Samples: U-251MG, SKOV3, A-549, Mouse brain.

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cst3
Gene Accession No. P01034, P14841, P21460
Gene Alias AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227
Gene Symbols Cst3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-146 of human Cystatin C (NP_0000901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13010, 1471, 25307
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.