Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin E/M/CST6 Antibody (037), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290741FR
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for ELISA.Specifications
Cystatin E/M/CST6 | |
Monoclonal | |
DyLight 680 | |
cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cystatin E/M/CST6 (NP_082899.1; Met1-Ala152). | |
0.1 mL | |
Cancer | |
1474 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
37 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction