Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin E/M/CST6 Antibody (037), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290741JF646
Description
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for ELISASpecifications
Cystatin E/M/CST6 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
037 | |
ELISA | |
cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cystatin E/M/CST6 (Accession#: NP_082899.1; Met1-Ala152). | |
0.1 mL | |
Cancer | |
1474 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction