Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin E/M/CST6 Antibody (RM0217-6F49), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212252PE
Description
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot, NeutralizationSpecifications
Cystatin E/M/CST6 | |
Monoclonal | |
PE | |
PBS | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Neutralization | |
RM0217-6F49 | |
Western Blot, Block/Neutralize | |
cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
Mouse recombinant Cystatin E/M/CST6 | |
0.1 mL | |
Cancer | |
1474 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction