Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin F Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298950F
Description
Cystatin F Polyclonal specifically detects Cystatin F in Human samples. It is validated for ELISA.Specifications
Cystatin F | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
8530 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
Produced in rabbits immunized with purified, recombinant Cystatin F (NP_003641.2; Met 1-His 145). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction