Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin F Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298244PE
Description
Cystatin F Polyclonal antibody specifically detects Cystatin F in Mouse samples. It is validated for ELISASpecifications
Cystatin F | |
Polyclonal | |
ELISA | |
CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
Produced in rabbits immunized with purified, recombinant Mouse Cystatin F (Uniprot#: O89098; Met1-Gln144) | |
0.1 mL | |
Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
8530 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction