Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cystatin M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547114
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547114 Supplier Invitrogen™ Supplier No. PA547114
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 25% cross-reactivity with recombinant mouse Cystatin E/M is observed and less than 1% cross-reactivity with recombinant human (rh) Cystatin A, rhCystatin B, rhCystatin C, rhCystatin D, rhCystatin S, rhCystatin SA, rhCystatin SN, rhFetuin A, and rhFetuin B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin M
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CST6
Gene Accession No. Q15828
Gene Alias 1110017E11Rik; AA960480; CST6; Cstb; cystatin 6; cystatin B; cystatin E/M; cystatin M; cystatin M/E; cystatin N; cystatin-6; Cystatin-B; cystatin-E; Cystatin-M; cysteine proteinase inhibitor; harlequin ichthyosis; harlequin ichthysosis; ichq; N28197; Stefin-B; Stfb
Gene Symbols CST6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cystatin E/M Arg29-Met149.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1474
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.