Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin S Antibody (OTI2H10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270481B
Description
Cystatin S Monoclonal specifically detects Cystatin S in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Cystatin S | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CST4 | |
Full length recombinant protein of human Cystatin S (NP_001890) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2H10 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1 | |
Mouse | |
Affinity Purified | |
Immunology | |
1472.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction