Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SA Antibody (005), Janelia Fluor™ 646 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289916JF646
Description
Cystatin SA Monoclonal antibody specifically detects Cystatin SA in Human samples. It is validated for ELISA, ELISASpecifications
Cystatin SA | |
Monoclonal | |
Janelia Fluor 646 | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin SA (NP_001313.1; Met1-Ala141). | |
0.1 mL | |
Cell Biology | |
1470 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
005 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction