Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SA Antibody (OTI1D6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270466C
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cystatin SA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CST2 | |
Full length recombinant protein of human Cystatin SA (NP_001313) produced in HEK293T cells. | |
0.1 mL | |
1470.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1D6 | |
Western Blot, Immunohistochemistry | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction