Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SA Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299787PCP
Description
Cystatin SA Polyclonal antibody specifically detects Cystatin SA in Human samples. It is validated for ELISASpecifications
Cystatin SA | |
Polyclonal | |
ELISA | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
Produced in rabbits immunized with purified, recombinant Human Cystatin SA (Accession#: NP_001313.1; Met 1-Ala 141) | |
0.1 mL | |
Cell Biology | |
1470 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction