Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SN Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299826JF549
Description
Cystatin SN Polyclonal antibody specifically detects Cystatin SN in Human samples. It is validated for ELISASpecifications
Cystatin SN | |
Polyclonal | |
ELISA | |
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
Produced in rabbits immunized with purified, recombinant Human Cystatin SN (Accession#: P01037-1; Met 1-Ser 141) | |
0.1 mL | |
Proteases & Other Enzymes | |
1469 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction