Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin SN Antibody (MM0234-5N51), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP212255AF532

Catalog No. NP2122A32


Only null left
Add to Cart

Description

Description

Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cystatin SN
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CST1
Human recombinant Cystatin SN
Proteases & Other Enzymes
1469
Store at 4°C in the dark.
Western Blot
MM0234-5N51
Western Blot
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family
Mouse
0.1 mL
Primary
Human
IgG2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.