Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SN Antibody (MM0234-5N51), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212255AF750
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot.Specifications
Cystatin SN | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CST1 | |
Human recombinant Cystatin SN | |
Proteases & Other Enzymes | |
1469 | |
Store at 4°C in the dark. |
Western Blot | |
MM0234-5N51 | |
Western Blot | |
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction