Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin/Stefin Homolog Antibody (RM0218-8D33), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212412AF647
Description
Cystatin/Stefin Homolog Monoclonal specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot.Specifications
Cystatin/Stefin Homolog | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CSTA | |
Mouse recombinant Cystatin/Stefin Homolog | |
0.1 mL | |
1475 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0218-8D33 | |
Western Blot | |
AREI, PSS4, STF1, STFA | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction