Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18601962
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

C1473, 7048-04-6, C3H7NO2S·HCl·H2O

Spectrum™ Chemical Cysteine Hydrochloride, Monohydrate, USP is a semi-essential amino acid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7048-04-6
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
IUPAC Name hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
SMILES [H+].O.[Cl-].N[C@@H](CS)C(O)=O

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade USP
Ignition Residue 0.4%
Loss on Drying 8.0-12%
Packaging Poly Bottle
Specific Rotation +5.7° to +6.8°
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.