Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cysteinyl Leukotriene R1/CysLTR1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.10 - $403.50
Specifications
Antigen | Cysteinyl Leukotriene R1/CysLTR1 |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cysteinyl Leukotriene R1/CysLTR1 Polyclonal antibody specifically detects Cysteinyl Leukotriene R1/CysLTR1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Cysteinyl Leukotriene R1/CysLTR1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Lipid and Metabolism | |
PBS with 50% glycerol, pH7.3. | |
10800 | |
IgG | |
Affinity purified |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
CysLT(1), CYSLT1CysLT1, CYSLT1R, CYSLTR, CysLTR1, Cysteinyl leukotriene D4 receptor, cysteinyl leukotriene receptor 1, G-protein coupled receptor HG55, HG55, HMTMF81, LTD4 receptor, MGC46139 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CYSLTR1 (NP_006630.1). MDETGNLTVSSATCHDTIDDFRNQVYSTLYSMISVVGFFGNGFVLYVLIKTYHKKSAFQVYMINLAVADLLCVCTLPLRVVYYVHKGIWLFGDFLCRLST | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title