Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cytarabine
GREENER_CHOICESDP

Catalog No. 452050 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-205-0 Supplier R&D Systems™ Supplier No. 4520/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Nucleoside analog; inhibits DNA replication

Nucleoside analog of deoxycytidine; inhibits DNA replication by incorporating into DNA (IC50 = 0.04 μM in L1210 and CEM cell lines). Displays no inhibitory effects on RNA synthesis. Causes S phase cell cycle arrest in ML-1 cell lines; cytotoxic in L5817Y leukemia cells. Antineoplastic and antileukemic agent.

Chemical Identifiers

CAS 147-94-4
Molecular Formula C9H13N3O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 243.22
Purity >99%
Molecular Weight (g/mol) 243.219
Chemical Name or Material Cytarabine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.