Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytidine 5'-Diphosphate Trisodium Salt Hydrate 95.0+%, TCI America™
SDP

Catalog No. C0523100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mg
100 mg
1 g

Chemical Identifiers

CAS 34393-59-4
Molecular Formula C9H12N3Na3O11P2
Molecular Weight (g/mol) 469.12
MDL Number MFCD00065201
InChI Key JENUKVZGXGULDX-LLWADOMFSA-K
Synonym cytidine 5'-diphosphate trisodium salt
PubChem CID 131674171
IUPAC Name trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 100 mg
Formula Weight 469.12
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine 5′-Diphosphate Trisodium Salt Hydrate
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.