Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cytidine, 99+%

Catalog No. AC111810500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111810500 50 g
AC111810100 10 g
AC111812500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111810500 Supplier Thermo Scientific Chemicals Supplier No. 111810500
Add to Cart

CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Absorbance 0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
Melting Point 210.0°C to 217.0°C
Color White
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 31,24
Merck Index 15,2783
Specific Rotation + 31.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alkalies,insoluble in organic solvents
Spectrophotometry 98 to 102% (lambda max. 280nm,pH 1) molar absorptivity 13000+/-2%,on dry basis
Transmittance 98% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 243.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, 99.5%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-610-9
RTECSNumber UW7370000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.