Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytidine 98.0+%, TCI America™
SDP

Catalog No. C052225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C052225G 25 g
C05221G 1 g
C05225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C052225G Supplier TCI America Supplier No. C052225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Color White
Quantity 25 g
Formula Weight 243.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine
EINECSNumber (5)-0941
RTECSNumber UW7370000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.