Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYTIP Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001002
Description
CYTIP Polyclonal specifically detects CYTIP in Human samples. It is validated for Western Blot, Peptide ELISA, Immunofluorescence.Specifications
CYTIP | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
B3-1, CASPCytohesin-associated scaffolding protein, Cbp HE, CYBRCytohesin-binding protein HE, CYTHIP, cytohesin 1 interacting protein, Cytohesin binder and regulator, cytohesin-1 interacting protein, cytohesin-interacting protein, HEPleckstrin homology Sec7 and coiled-coil domains-binding protein, pleckstrin homology, Sec7 and coiled/coil domains, binding protein, pleckstrin homology, Sec7 and coiled-coil domains, binding protein, PSCDBPcytohesin binding protein HE | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Pig, Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunofluorescence | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/mL, Peptide ELISA Detection limit 1:16000, Immunofluorescence 10 ug/mL | |
NP_004279. | |
CYTIP | |
Peptide with sequence SLQRLLQHSSNGN-C corresponding to N-Terminus according to NP_004279.3. | |
0.1 mg | |
Signal Transduction | |
9595 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction