Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYTIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552400 Supplier Invitrogen™ Supplier No. PA552400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAYSSYSTLT GSLTMDDNRR IQMLADTVAT LPRGRKQLAL TRSSSLSDFS WSQRKLVTVE KQDNETFGFE IQSYRPQNQN ACSSEMFTLI CKIQEDSPAH CAGLQAGDVL ANINGVSTEG F Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYTIP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYTIP
Gene Accession No. O60759
Gene Alias A130053M09Rik; AI462064; B3-1; C80816; CASP; Cbp; cbp HE; CYBR; CYTHIP; Cytip; cytohesin 1 interacting protein; cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; cytohesin-interacting protein; cytohesin-interacting protein-like; HE; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; Pscdbp; Sec7; tamalin; zmp:0000001116
Gene Symbols CYTIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYTIP. Recombinant protein control fragment (Product #RP-89673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9595
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.