Learn More
Invitrogen™ CYTIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114192
Description
Immunogen sequence: QLALTRSSSL SDFSWSQRKL VTVEKQDNET FGFEIQSYRP QNQNACSSEM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Guinea Pig: 75%; Human: 100%; Mouse: 79%; Pig: 79%; Rat: 79%.
The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.
Specifications
CYTIP | |
Polyclonal | |
Unconjugated | |
CYTIP | |
A130053M09Rik; AI462064; B3-1; C80816; CASP; Cbp; cbp HE; CYBR; CYTHIP; Cytip; cytohesin 1 interacting protein; cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; cytohesin-interacting protein; cytohesin-interacting protein-like; HE; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; Pscdbp; Sec7; tamalin; zmp:0000001116 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9595 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60759 | |
CYTIP | |
Immunogen sequence: QLALTRSSSL SDFSWSQRKL VTVEKQDNET FGFEIQSYRP QNQNACSSEM (aa 58-107). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.