Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYTIP Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518019
Description
This antibody is predicted to react with canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.
Specifications
CYTIP | |
Polyclonal | |
Unconjugated | |
CYTIP | |
A130053M09Rik; AI462064; B3-1; C80816; CASP; Cbp; cbp HE; CYBR; CYTHIP; Cytip; cytohesin 1 interacting protein; cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; cytohesin-interacting protein; cytohesin-interacting protein-like; HE; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; Pscdbp; Sec7; tamalin; zmp:0000001116 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
9595 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O60759 | |
CYTIP | |
Synthetic peptide sequence (SLQRLLQHSSNGN) corresponding to the N-terminus amino acids of CYTIP (aa 2-15). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction