Learn More
Cytiva Cytiva Dissociation Enzyme Mix A - Lung
Catalog No. 09924465 Shop All Cytiva Products
Change view
Click to view available options
Form:
Lyophilized
Catalog No. Form
09-924-465 Lyophilized
1 options
Catalog No. 09-924-465 Supplier Cytiva Supplier No. 29740243
Get an Extra Life Sciences Product with Purchase
View all promotions
Add to Cart
Add to Cart
Enzymatic mixes for lung tissue optimized for manual and semi-automated dissociation in single-cell workflows.
  • Dissociation enzyme mixes (for research only) for lung are kits intended for the dissociation of lung tissues in single-cell sample preparation workflows
  • The kits have been optimized for use with manual and semi-automated tissue disaggregation techniques
  • Work effectively at either 4°C or 37°C
  • Improved single cell viability and reduced cellular aggregation
  • Ready-made solution for lung tissue - Optimized for the dissociation of the different segments that are operative in respective tissue
  • Optimized for use with manual tissue disaggregation techniques and semi-automated techniques including the VIA Extractor™ tissue disaggregator
  • Performance data is not available for Enzyme mix A (for lung) using semi-automated methods at temperatures of 37°C
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Product Type Cell Culture Dissociation Reagent
Sterility Non-sterile
With Additives None
Without Additives None