Learn More
Cytiva Cytiva Dissociation Enzyme Mix B - Liver
Catalog No. 09924466 Shop All Cytiva Products
Change view
Click to view available options
Form:
Lyophilized
Catalog No. Form
09-924-466 Lyophilized
1 options
Catalog No. 09-924-466 Supplier Cytiva Supplier No. 29751004
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Enzymatic mixes for liver tissue optimized for manual and semi-automated dissociation in single-cell workflows.
  • Dissociation enzyme mixes (for research only) for liver are kits intended for the dissociation of liver tissues in single-cell sample preparation workflows
  • The kits have been optimized for use with manual and semi-automated tissue disaggregation techniques
  • Work effectively at 37°C
  • Improved single cell viability and reduced cellular aggregation
  • Ready-made solution for liver tissue - Optimized for the dissociation of the different segments that are operative in respective tissue
  • Optimized for use with manual tissue disaggregation techniques and semi-automated techniques including the VIA Extractor™ tissue disaggregator
  • Enzyme mix B (for liver) is recommended for use at 37°C as liver is very sensitive to cold
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Product Type Cell Culture Dissociation Reagent
Sterility Non-sterile
With Additives None
Without Additives None