Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Cyto-Chex™ BCT Storage and Transport Reagent
SDP

Catalog No. p-3143300
Click to view available options
Quantity:
6 x 5 mL
25 x 5 mL

Cyto-Chex BCT is a direct draw blood collection tube that maintains cellular morphology and surface antigen expression, including cluster of differentiation (CD) markers, prior to analysis by flow cytometry. Whole blood samples collected in Cyto-Chex BCT can be stored at room temperature for up to 14 days. Cyto-Chex BCT has been FDA 510(k) cleared for consistent recovery of HIV-associated lymphocyte subsets for up to 14 days.

  • Stabilizes whole blood samples for up to 14 days prior to analysis by flow cytometry.
  • FDA 510(k) cleared for consistent recovery of HIV-associated lymphocyte subsets (CD3, CD4, CD8, CD19, CD16/CD56 and CD45) for up to 14 days of room temperature storage.
  • Available in 5 mL and 2 mL vials.
  • Limits the adverse effects of time, storage and transport conditions on sample integrity.
  • Reduces the need for immediate sample processing and analysis.
  • Allows for the batching of samples, optimizing laboratory workflow.

Specifications

Certifications/Compliance CE
Content And Storage 18°C to 30°C
Format Tube
Type Cyto-Chex™ BCT 7 Day Immunophenotyping Preservative
Includes Cyto-Chex™ BCT contains the anticoagulant, K3EDTA, and a cell preservative in a liquid medium
Intended Use Collection, transport, and retention of Whole Blood samples for immunophenotyping
Quantity 6 x 5 mL
Stability 7 Days
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.