Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cyto-Plex™ Carboxylated Particles

Catalog No. 09980701
Click to view available options
Diameter (Metric):
4 μm
5 μm
Quantity:
1 mL
Level:
Level 1
Level 11
Level 4
Level 6
Level 8
Contains sodium azide.
Contains sodium azide.

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters

Recommended for:

  • Open platform microspheres for suspension array analysis in flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Additive Sodium Azide Preservative, 0.05% Tween-20
Content And Storage Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.
Contents Polystyrene microspheres in water
Description Cyto-Plex Carboxylated Particles, 4µm
Color Red
Material Polystyrene with Copolymer Grafted Surface
Product Type Carboxylated Particle
Dye Type Firefli™ Fluorescent Red (635/700nm)
Includes Material Safety Data Sheet (MSDS) and Package Insert Sheet
Level Level 1
Particle Density 1.06g/cm³
Quantity 1 mL
Stability Product stable for a minimum of 12 months
Surface Modification Carboxylate-Modified
Uniformity ≤2% CV
Diameter (Metric) 4 μm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.