Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome b reductase 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | Cytochrome b reductase 1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cytochrome b reductase 1 Polyclonal specifically detects Cytochrome b reductase 1 in Human samples. It is validated for Western Blot.Specifications
| Cytochrome b reductase 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| cytochrome b reductase 1, DCYTBFRRS3FLJ23462, Duodenal cytochrome b, Ferric-chelate reductase 3 | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human Cytochrome b reductase 1 (NP_001120855.1). Peptide sequence PNGGTEQGARGSMPAYSGNNMDKSDSELNSEVAARKRNLALDEAGQRSTM | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 79901 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title