Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome b245 alpha Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Cytochrome b245 alpha
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094004
SDP
View Documents
Novus Biologicals
NBP30340320UL
20 μL
Each for $181.90
Only null left
Add to Cart
 
NB094003
SDP
View Documents
Novus Biologicals
NBP303403100UL
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Cytochrome b245 alpha Polyclonal specifically detects Cytochrome b245 alpha in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytochrome b245 alpha
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
1535
IgG
Affinity purified
Immunofluorescence
Unconjugated
Rabbit
Human
cytochrome b light chain, Cytochrome b(558) alpha chain, cytochrome b(558) alpha-subunit, cytochrome b, alpha polypeptide, cytochrome b-245 light chain, cytochrome b-245, alpha polypeptide, Cytochrome b558 subunit alpha, flavocytochrome b-558 alpha polypeptide, Neutrophil cytochrome b 22 kDa polypeptide, p22 phagocyte B-cytochrome, p22phox, p22-phox, Superoxide-generating NADPH oxidase light chain subunit
Recombinant fusion protein containing a sequence corresponding to amino acids 126-195 of human Cytochrome b245 alpha (NP_000092.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.